missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Isoserine, 98%

Manufacturer:  Alfa Aesar™ H5341203

 View more versions of this product

Catalog No. AAH5341203



Description

Description

Applications
DL-Isoserine has been used in the synthesis of L-isoserine derivatives s as aminopeptidase N inhibitors, Syntheses of (2R,3S)-3-Hydroxyleucinate.

Solubility
Soluble in HCl.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

DL-Isoserine
C3H7NO3
1721413
BMYNFMYTOJXKLE-UHFFFAOYSA-N
3-amino-2-hydroxypropanoic acid
11267
98%
1g
565-71-9
MFCD00008138
dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947, dl-isoserine
C(C(C(=O)O)O)N
105.093
105.09
235°C

Chemical Identifiers

565-71-9
105.093
BMYNFMYTOJXKLE-UHFFFAOYSA-N
11267
C(C(C(=O)O)O)N
C3H7NO3
MFCD00008138
dl-isoserine, isoserine, 3-amino-2-hydroxypropionic acid, propanoic acid, 3-amino-2-hydroxy, 2-hydroxy-3-aminopropanoic acid, h-dl-isoser-oh, inverted exclamation marka-3-amino-2-hydroxypropionic acid, rs-isoserine, pubchem23947, dl-isoserine
3-amino-2-hydroxypropanoic acid
SDS Documents