Please login to your online account to display your discounted pricing

DL-Isoproterenol Sulfate Dihydrate 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  146190250

 View more versions of this product

Catalog No. AC146190250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (on dry substance)
Physical Form Fine Crystalline Powder
Formula Weight 556.64
Quantity 25g
Color White
Merck Index 15, 5263
Synonym N-Isopropyl-DL-noradrenaline sulfate dihydrate
Packaging Glass bottle
Water 5 to 7.5%
Chemical Name or Material DL-Isoproterenol sulfate dihydrate
Name Note 97%
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula 5 H2SO4·H2O
Linear Formula 3, 4-(HO)2C6H3CH(OH)CH2NHCH(CH3)2.H2SO4
MDL Number MFCD07371352
CAS 6700-39-6

Specifications

Chemical Name or Material DL-Isoproterenol sulfate dihydrate
Name Note 97%
CAS 6700-39-6
Assay ≥96.0% (on dry substance)
Assay Percent Range 97%
Molecular Formula 5 H2SO4·H2O
Linear Formula 3, 4-(HO)2C6H3CH(OH)CH2NHCH(CH3)2.H2SO4
Formula Weight 556.64
MDL Number MFCD07371352
Synonym N-Isopropyl-DL-noradrenaline sulfate dihydrate