Please login to your online account to display your discounted pricing

DL-Isoproterenol Sulfate Dihydrate 97%, ACROS Organics™

$119.05 - $119.05

Chemical Identifiers

Chemical Name or Material DL-Isoproterenol sulfate dihydrate
Name Note 97%
CAS 6700-39-6
Assay 96.0% min. (on dry substance)
Assay Percent Range 97%
Molecular Formula 5 H2SO4·H2O
Linear Formula 3, 4-(HO)2C6H3CH(OH)CH2NHCH(CH3)2.H2SO4
Formula Weight 556.64g/mol
MDL Number MFCD07371352
Synonym N-Isopropyl-DL-noradrenaline sulfate dihydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146190250

  SDS
thermo fisher scientific
146190250
Glass bottle 25g Each for $119.05
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Water 5 to 7.5%
Color White
Quantity 25g
Chemical Name or Material DL-Isoproterenol sulfate dihydrate
Name Note 97%
CAS 6700-39-6
Physical Form White fine crystalline powder
Assay 96.0% min. (on dry substance)
Assay Percent Range 97%
Molecular Formula 5 H2SO4·H2O
Linear Formula 3, 4-(HO)2C6H3CH(OH)CH2NHCH(CH3)2.H2SO4
Formula Weight 556.64g/mol
MDL Number MFCD07371352
Merck Index 15, 5263
Synonym N-Isopropyl-DL-noradrenaline sulfate dihydrate