Please login to your online account to display your discounted pricing

DL-Isoleucine, ≥99%, MP Biomedicals™


Quantity 10g

Manufacturer: mp biomedicals  10208210

 View more versions of this product

Catalog No. ICN10208210

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 0.991
Formula Weight 131.17
Physical Form Powder
Color White
Quantity 10g
EINECS Number 207-139-8
Recommended Storage Store at Room Temperature (15 to 30°C).
Assay Percent Range ≥99%
CAS 443-79-8
Molecular Formula C6H13NO2
Chemical Name or Material DL-Isoleucine
Synonym 2-Amino-3-Methylpentanoic Acid
MDL Number MFCD00004268

Specifications

Chemical Name or Material DL-Isoleucine
CAS 443-79-8
Physical Form Powder
Assay Percent Range ≥99%
Molecular Formula C6H13NO2
Formula Weight 131.17
MDL Number MFCD00004268
Synonym 2-Amino-3-Methylpentanoic Acid
Accessories

Store at Room Temperature (15 to 30°C).