Please login to your online account to display your discounted pricing

DL-Isoleucine, ≥99%, MP Biomedicals™

$58.81 - $58.81

Chemical Identifiers

Chemical Name or Material DL-Isoleucine
CAS 443-79-8
Physical Form Powder
Assay Percent Range ≥99%
Molecular Formula C6H13NO2
Formula Weight 131.17
MDL Number MFCD00004268
Synonym 2-Amino-3-Methylpentanoic Acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN10208210

 
mp biomedicals
10208210
10g Each for $58.81
Description & Specifications

Specifications

Chemical Name or Material DL-Isoleucine
CAS 443-79-8
Recommended Storage Store at Room Temperature (15 to 30°C).
EINECS Number 207-139-8
Physical Form Powder
Assay 0.991
Assay Percent Range ≥99%
Molecular Formula C6H13NO2
Formula Weight 131.17
MDL Number MFCD00004268
Synonym 2-Amino-3-Methylpentanoic Acid
Color White
Quantity 10g
Safety and Handling

Store at Room Temperature (15 to 30°C).