missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Isocitric acid, trisodium salt hydrate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC205010050

 View more versions of this product

Catalog No. AC205010050



Specifications

Specifications

DL-Isocitric acid, trisodium salt hydrate
Glass bottle
White
1637-73-6
C6H5Na3O7·xH2O
MFCD00149284
dl-isocitric acid, dl-isocitric acid trisodium salt, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid, trisodium salt hydrate, isocitric acid trisodium salt, isocitric acid, dl, sodium isocitrate, tripotassium isocitrate, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate
HWMVXEKEEAIYGB-UHFFFAOYSA-K
trisodium;1-hydroxypropane-1,2,3-tricarboxylate
168942
Crystalline Powder
Authentic
18.5% max. (K.F.)
5g
95%
NaO2CCH2CH(CO2Na)CH(OH)CO2Na·xH2O
03, 555
Solubility in water: soluble in water
C(C(C(C(=O)[O-])O)C(=O)[O-])C(=O)[O-].[Na+].[Na+].[Na+]
258.07
258.07
95%

Chemical Identifiers

1637-73-6
258.07
HWMVXEKEEAIYGB-UHFFFAOYSA-K
168942
C(C(C(C(=O)[O-])O)C(=O)[O-])C(=O)[O-].[Na+].[Na+].[Na+]
C6H5Na3O7·xH2O
MFCD00149284
dl-isocitric acid, dl-isocitric acid trisodium salt, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid, trisodium salt hydrate, isocitric acid trisodium salt, isocitric acid, dl, sodium isocitrate, tripotassium isocitrate, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate
trisodium;1-hydroxypropane-1,2,3-tricarboxylate
Safety and Handling

Safety and Handling

EINECSNumber : 216-668-3

SDS Documents