missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Indole-3-lactic acid, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC122222500

 View more versions of this product

Catalog No. AC122222500



Specifications

Specifications

DL-Indole-3-lactic acid
Glass bottle
145°C to 147°C
832-97-3
MFCD00005642
1-alpha-hydroxy-1h-indole-3-propionic acid, 2-hydroxy-3-1h-indol-3-yl propanoic acid, 2-hydroxy-3-indol-3-ylpropanoic acid, 3-indol-3-yl lactic acid, 3-indolelactic acid, alpha-hydroxy-1h-indole-3-propanoic acid, dl-indole-3-lactic acid, indole-3-lactic acid, indolelactate, indolelactic acid
XGILAAMKEQUXLS-UHFFFAOYSA-N
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
92904
205.21
≥97.5%
Authentic
Brown to White
250mg
C11H11NO3
22, I, 553
Solubility in water: insoluble
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
205.21
CHEBI:24813
Crystalline Powder

Chemical Identifiers

832-97-3
205.21
XGILAAMKEQUXLS-UHFFFAOYSA-N
92904
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
C11H11NO3
MFCD00005642
1-alpha-hydroxy-1h-indole-3-propionic acid, 2-hydroxy-3-1h-indol-3-yl propanoic acid, 2-hydroxy-3-indol-3-ylpropanoic acid, 3-indol-3-yl lactic acid, 3-indolelactic acid, alpha-hydroxy-1h-indole-3-propanoic acid, dl-indole-3-lactic acid, indole-3-lactic acid, indolelactate, indolelactic acid
CHEBI:24813
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 212-627-9

SDS Documents