missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Indole-3-lactic acid, ACROS Organics™

$100.83 - $270.96

Chemical Identifiers

CAS 832-97-3
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00005642
InChI Key XGILAAMKEQUXLS-UHFFFAOYSA-N
Synonym 1-alpha-hydroxy-1h-indole-3-propionic acid, 2-hydroxy-3-1h-indol-3-yl propanoic acid, 2-hydroxy-3-indol-3-ylpropanoic acid, 3-indol-3-yl lactic acid, 3-indolelactic acid, alpha-hydroxy-1h-indole-3-propanoic acid, dl-indole-3-lactic acid, indole-3-lactic acid, indolelactate, indolelactic acid
PubChem CID 92904
ChEBI CHEBI:24813
IUPAC Name 2-hydroxy-3-(1H-indol-3-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122222500
SDS View Documents
Thermo Fisher Scientific
AC122222500
250mg Glass bottle
Each for $100.83
 
AC122220010
SDS View Documents
Thermo Fisher Scientific
AC122220010
1g Glass bottle
Each for $270.96
 
Description

Description

Specifications

Chemical Identifiers

832-97-3
205.21
XGILAAMKEQUXLS-UHFFFAOYSA-N
92904
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
C11H11NO3
MFCD00005642
1-alpha-hydroxy-1h-indole-3-propionic acid, 2-hydroxy-3-1h-indol-3-yl propanoic acid, 2-hydroxy-3-indol-3-ylpropanoic acid, 3-indol-3-yl lactic acid, 3-indolelactic acid, alpha-hydroxy-1h-indole-3-propanoic acid, dl-indole-3-lactic acid, indole-3-lactic acid, indolelactate, indolelactic acid
CHEBI:24813
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O

Specifications

Authentic
Brown to White
250mg
C11H11NO3
22, I, 553
Solubility in water: insoluble
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
205.21
CHEBI:24813
Crystalline Powder
DL-Indole-3-lactic acid
Glass bottle
145°C to 147°C
832-97-3
MFCD00005642
1-alpha-hydroxy-1h-indole-3-propionic acid, 2-hydroxy-3-1h-indol-3-yl propanoic acid, 2-hydroxy-3-indol-3-ylpropanoic acid, 3-indol-3-yl lactic acid, 3-indolelactic acid, alpha-hydroxy-1h-indole-3-propanoic acid, dl-indole-3-lactic acid, indole-3-lactic acid, indolelactate, indolelactic acid
XGILAAMKEQUXLS-UHFFFAOYSA-N
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
92904
205.21
≥97.5%
Safety and Handling

Safety and Handling

EINECSNumber : 212-627-9

SDS