Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Homocysteic acid, 98%

$53.88 - $53.88

Chemical Identifiers

Chemical Name or Material DL-Homocysteic acid
CAS 504-33-6
Molecular Formula C4H9NO5S
Formula Weight 183.18
MDL Number MFCD00007536
Beilstein 1783391
Synonym 2-Amino-4-sulfobutyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0912203

 
alfa aesar™
L0912203
1g Each for $53.88
Description & Specifications

Specifications

Formula Weight 183.18
Molecular Formula C4H9NO5S
Assay 98%
MDL Number MFCD00007536
CAS 504-33-6
Beilstein 1783391
Synonym 2-Amino-4-sulfobutyric acid
Melting Point ≈273°C
Chemical Name or Material DL-Homocysteic acid
Odor Odorless
Quantity 1g