missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Ethyl 2-bromopropionate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 118142500

 View more versions of this product

Catalog No. AC118142500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

DL-Ethyl 2-bromopropionate
51°C
Glass bottle
1.394
Colorless to Yellow
99%
98.5% min. (GC)
CH3CH(Br)CO2C2H5
02,255
2-bromopropanoic acid ethyl ester
ARFLASKVLJTEJD-UHFFFAOYSA-N
ethyl 2-bromopropanoate
79040
Liquid
1.3940g/mL
Authentic
1.4450 to 1.447
156°C to 160°C
250g
535-11-5
C5H9BrO2
MFCD00000144
15,3827
Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
CCOC(=O)C(C)Br
181.03
181.03
99%

Chemical Identifiers

535-11-5
181.03
ARFLASKVLJTEJD-UHFFFAOYSA-N
79040
CCOC(=O)C(C)Br
C5H9BrO2
MFCD00000144
2-bromopropanoic acid ethyl ester
ethyl 2-bromopropanoate
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.<

GHS Signal Word: Danger

EINECSNumber : 208-609-5

RTECSNumber : UA2451730

TSCA : TSCA

SDS Documents