missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Ethionine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 146170025

 View more versions of this product

Catalog No. AC146170025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

265°C to 269°C
67-21-0
98%
Authentic
MFCD00063102
Glass Bottle
GGLZPLKKBSSKCX-UHFFFAOYNA-N
2-amino-4-ethylsulfanylbutanoic acid
6205
163.23
2.5 g
DL-Ethionine
White
C6H13NO2S
C2H5SCH2CH2CH(NH2)CO2H
15, 3792
dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
CCSCCC(N)C(O)=O
163.24
CHEBI:68662
≥97.5%
Crystalline Powder or Flakes

Chemical Identifiers

67-21-0
163.24
GGLZPLKKBSSKCX-UHFFFAOYNA-N
6205
2-amino-4-ethylsulfanylbutanoic acid
C6H13NO2S
MFCD00063102
dl-ethionine, s-ethylhomocysteine, aethionin, s-ethyl-dl-homocysteine, 2-amino-4-ethylthio butyric acid, dl-homocysteine, s-ethyl, ethionine, dl, +--ethionine, homocysteine, s-ethyl, 2-amino-4-ethylthio butanoic acid
CHEBI:68662
CCSCCC(N)C(O)=O
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only