missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-erythro-Dihydrosphingosine

$789.57 - $789.57

Chemical Identifiers

CAS 3102-56-5
Molecular Formula C18H39NO2
Molecular Weight (g/mol) 301.515
MDL Number MFCD00079141
InChI Key OTKJDMGTUTTYMP-MSOLQXFVSA-N
Synonym dl-erythro-dihydrosphingosine, dl-sphinganine, 2r,3s-2-aminooctadecane-1,3-diol, dl-erythro-1,3-dihydroxy-2-aminooctadecane, lopac-d-6908, l-erythro-dihydrosphingosine, dl-erythro-dihydrosphingosine, synthetic
PubChem CID 6603822
IUPAC Name (2R,3S)-2-aminooctadecane-1,3-diol
SMILES CCCCCCCCCCCCCCCC(C(CO)N)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62103MF View Documents Alfa Aesar™
J62103MF
50mg Each for $789.57
 
Description

Description

Applications
DL-erythro-Dihydrosphingosine is an inhibitor of PKC (protein kinase C). This compound also blocks PLA2 (phospholipases A2) and the D-sphingosine precursor.

Solubility
Soluble in water (partly miscible), chloroform, 100% warm ethanol (25 mg/ml), warm DMSO (25 mg/ml), and methanol.

Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at -20°C.
Specifications

Chemical Identifiers

3102-56-5
301.515
OTKJDMGTUTTYMP-MSOLQXFVSA-N
6603822
CCCCCCCCCCCCCCCC(C(CO)N)O
C18H39NO2
MFCD00079141
dl-erythro-dihydrosphingosine, dl-sphinganine, 2r,3s-2-aminooctadecane-1,3-diol, dl-erythro-1,3-dihydroxy-2-aminooctadecane, lopac-d-6908, l-erythro-dihydrosphingosine, dl-erythro-dihydrosphingosine, synthetic
(2R,3S)-2-aminooctadecane-1,3-diol

Specifications

White
3102-56-5
MFCD00079141
dl-erythro-dihydrosphingosine, dl-sphinganine, 2r,3s-2-aminooctadecane-1,3-diol, dl-erythro-1,3-dihydroxy-2-aminooctadecane, lopac-d-6908, l-erythro-dihydrosphingosine, dl-erythro-dihydrosphingosine, synthetic
CCCCCCCCCCCCCCCC(C(CO)N)O
301.515
301.51
DL-erythro-Dihydrosphingosine
50mg
C18H39NO2
1724234
OTKJDMGTUTTYMP-MSOLQXFVSA-N
(2R,3S)-2-aminooctadecane-1,3-diol
6603822
Powder
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C