missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Dithiothreitol, ≥99.0%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210059725
Specifications
DL-Dithiothreitol | |
White | |
C4H10O2S2 | |
1, 4-Dithio-DL-threitol, Cleland Reagent, DTT | |
VHJLVAABSRFDPM-UHFFFAOYNA-N | |
1,4-disulfanylbutane-2,3-diol | |
154.25 |
27565-41-9 | |
≥99.0% | |
MFCD00004877,MFCD00064305 | |
25g | |
OC(CS)C(O)CS | |
154.24 | |
Powder |
Chemical Identifiers
27565-41-9 | |
154.24 | |
VHJLVAABSRFDPM-UHFFFAOYNA-N | |
1,4-disulfanylbutane-2,3-diol |
C4H10O2S2 | |
MFCD00004877,MFCD00064305 | |
1, 4-Dithio-DL-threitol, Cleland Reagent, DTT | |
OC(CS)C(O)CS |
Safety and Handling
EINECSNumber : 248-531-9