missing translation for 'onlineSavingsMsg'
Learn More

DL-Cystine

CAS: 923-32-0 | C6H12N2O4S2 | 240.292 g/mol

Supplier:  Thermo Scientific Chemicals J6356406

 View more versions of this product

Catalog No. AAJ6356406


Only null left
Explore available promotions

Description

Description

DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-cystine is used in the preparation of sulfur-containing dimeric and monomeric surfactants.

Solubility
Soluble in dilute acid and alkali solution, highly soluble in water, not soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

227°C (decomposition)
923-32-0
C6H12N2O4S2
1728095
dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l
LEVWYRKDKASIDU-UHFFFAOYSA-N
2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid
240.292
CHEBI:17376
Powder
DL-Cystine
White
MFCD00084652
14,2782
Soluble in dilute acid and alkali solution,highly soluble in water,not soluble in alcohol.
C(C(C(=O)O)N)SSCC(C(=O)O)N
5 g
595
240.3

Chemical Identifiers

923-32-0
240.292
LEVWYRKDKASIDU-UHFFFAOYSA-N
595
2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid
C6H12N2O4S2
MFCD00084652
dl-cystine, cystine, d-cystine, 3,3'-disulfanediylbis 2-aminopropanoic acid, dicysteine, cystin, h-dl-cys-oh 2, .beta.,.beta.'-dithiodialanine, cystine, dl, cystine, l
CHEBI:17376
C(C(C(=O)O)N)SSCC(C(=O)O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 213-094-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only