missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Cysteine hydrochloride, 95%, Acros Organics™

Manufacturer:  Acros Organics AC448260250

 View more versions of this product

Catalog No. AC448260250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

DL-Cysteine hydrochloride
21.49 to 24.23% (C) 8.35 to 9.42% (N)
HSCH2CH(NH2)CO2H·HCl
1% max. Vacuum, room temp, 20 hours+
Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
C(C(C(=O)O)N)S.Cl
157.612
157.61
156.0°C to 161.0°C
10318-18-0
C3H8ClNO2S
Authentic
dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
IFQSXNOEEPCSLW-UHFFFAOYSA-N
2-amino-3-sulfanylpropanoic acid;hydrochloride
25150
≥94.0% (HPLC)
25g

Chemical Identifiers

10318-18-0
157.612
dl-cysteine hydrochloride, 2-amino-3-mercaptopropanoic acid hydrochloride, cysteine hydrochloride, +--cysteine hydrochloride, dl-cysteine hcl, h-dl-cys-oh.hcl, cysteine, hydrochloride, dl, dl-cysteine hydrochloride hydrate, dl-cysteine hydrochloride anhydrous, 2-amino-3-sulfanylpropanoic acid hydrochloride
2-amino-3-sulfanylpropanoic acid;hydrochloride
C3H8ClNO2S
IFQSXNOEEPCSLW-UHFFFAOYSA-N
25150
C(C(C(=O)O)N)S.Cl
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

Warning

SDS Documents