missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Cysteine, 96%

Manufacturer:  Alfa Aesar™ H5612606

 View more versions of this product

Catalog No. AAH5612606



Description

Description

Applications
DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.

Solubility
Soluble in water (partly), 1 N HCl (25 mg/ml), ethanol, and acetic acid. Insoluble in acetone.

Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

DL-Cysteine
C3H7NO2S
1721406
Soluble in water (partly),1 N HCl (25mg/ml),ethanol,and acetic acid. Insoluble in acetone.
C(C(C(=O)O)N)S
121.154
CHEBI:15356
96%
5g
3374-22-9
MFCD00004881
+--cysteine
XUJNEKJLAYXESH-UHFFFAOYSA-N
2-amino-3-sulfanylpropanoic acid
594
121.16
225°C (decomposition)

Chemical Identifiers

3374-22-9
121.154
XUJNEKJLAYXESH-UHFFFAOYSA-N
594
2-amino-3-sulfanylpropanoic acid
C3H7NO2S
MFCD00004881
+--cysteine
CHEBI:15356
C(C(C(=O)O)N)S
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 222-160-2

RTECSNumber : HA1595000

TSCA : No

Recommended Storage : Ambient temperatures

Documents