Learn More
DL-Cysteine, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5612622
Description
DL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Cysteine is a NMDA agonist at quisqualate receptors at high concentration.
Solubility
Soluble in water (partly), 1 N HCl (25 mg/ml), ethanol, and acetic acid. Insoluble in acetone.
Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
225°C (decomposition) | |
3374-22-9 | |
MFCD00004881 | |
dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh | |
XUJNEKJLAYXESH-UHFFFAOYNA-N | |
2-amino-3-sulfanylpropanoic acid | |
594 | |
121.16 | |
100 g |
DL-Cysteine | |
C3H7NO2S | |
1721406 | |
Soluble in water (partly),1 N HCl (25mg/ml),ethanol,and acetic acid. Insoluble in acetone. | |
NC(CS)C(O)=O | |
121.15 | |
CHEBI:15356 | |
96% |
Chemical Identifiers
3374-22-9 | |
121.15 | |
XUJNEKJLAYXESH-UHFFFAOYNA-N | |
594 | |
2-amino-3-sulfanylpropanoic acid |
C3H7NO2S | |
MFCD00004881 | |
dl-cysteine, cysteine, 2-amino-3-mercaptopropanoic acid, dl-cystein, cysteine, dl, +--cysteine, dl-cysteina polish, h-dl-cys-oh, 2-amino-3-mercaptopropionic acid, l-hsch2ch nh2 cooh | |
CHEBI:15356 | |
NC(CS)C(O)=O |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 222-160-2
RTECSNumber : HA1595000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only