missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Citrulline, 97%

Manufacturer:  Alfa Aesar™ A1138622

 View more versions of this product

Catalog No. AAA1138622



Specifications

Specifications

DL-Citrulline
627-77-0
C6H13N3O3
DL-2-Amino-5-ureidovaleric acid
C(CC(C(=O)O)N)CNC(=O)N
175.188
CHEBI:18211
100g
97%
MFCD00007955
RHGKLRLOHDJJDR-UHFFFAOYSA-N
2-amino-5-(carbamoylamino)pentanoic acid
833
175.19

Chemical Identifiers

627-77-0
175.188
RHGKLRLOHDJJDR-UHFFFAOYSA-N
833
2-amino-5-(carbamoylamino)pentanoic acid
C6H13N3O3
MFCD00007955
DL-2-Amino-5-ureidovaleric acid
CHEBI:18211
C(CC(C(=O)O)N)CNC(=O)N
SDS Documents