Please login to your online account to display your discounted pricing

DL-Carnitine Hydrochloride 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  108451000

 View more versions of this product

Catalog No. AC108451000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder, Crystals, Needles and/or Chunks
Assay ≥98.5% (total base)
Formula Weight 197.66
Quantity 100g
Color White to almost white
Merck Index 15, 1849
Melting Point 194° to 200°C
pH 1.8 to 2.5 (10% soln.)
EINECS Number 207-309-1
TSCA TSCA
RTECS Number BP2979140
Packaging Plastic bottle
Specific Rotation −1° to +1° (20°C, 589nm) (c=1, H2O)
Synonym 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride
Health Hazard 1 Danger
Water 2% max.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material DL-Carnitine hydrochloride
Molecular Formula C7H15NO3·HCl
Linear Formula HO2CCH2CH(OH)CH2N(CH3)3Cl
MDL Number MFCD00011904
CAS 461-05-2
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Ammonium 500ppm max.
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell

Specifications

Chemical Name or Material DL-Carnitine hydrochloride
Name Note 99%
CAS 461-05-2
Assay ≥98.5% (total base)
Assay Percent Range 99%
Molecular Formula C7H15NO3·HCl
Linear Formula HO2CCH2CH(OH)CH2N(CH3)3Cl
Formula Weight 197.66
MDL Number MFCD00011904
Synonym 3-Hydroxy-4-(trimethylammonio)butanoate hydrochloride
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

SDS & Safety Documents