missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ DL-Asparagine monohydrate, 98%
Supplier: Thermo Scientific™ 167121000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
DL-Asparagine monohydrate | |
3130-87-8 | |
C4H8N2O3 | |
H2NCOCH2CH(NH2)CO2H·H2O | |
04, 484 | |
Plastic bottle | |
DCXYFEDJOCDNAF-UHFFFAOYNA-N | |
2,4-diamino-4-oxobutanoic acid;hydrate | |
522362 | |
150.14 | |
10.5 to 13.5% (K.F.) |
98% | |
98% | |
Authentic | |
MFCD00151039,MFCD00008036,MFCD00064400 | |
14, 837 | |
dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina | |
NC(CC(N)=O)C(O)=O | |
132.12 | |
90% min. (c=10, 2N HCl, 430nm) 1cm cell | |
≥97.5% | |
100g |
Safety and Handling
EINECSNumber : 221-521-1
TSCA : TSCA
RUO â Research Use Only