Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Arginine monohydrochloride monohydrate, 98+%

$35.55 - $35.55

Chemical Identifiers

Chemical Name or Material DL-Arginine monohydrochloride monohydrate
CAS 332360-01-7
Molecular Formula C6H14N4O2·HCl·H2O
Formula Weight 228.69 (210.67 Anhydrous)
MDL Number MFCD00151031
Beilstein 4158960
Synonym DL-2-Amino-5-guanidinopentanoic acid monohydrochloride monohydrate; H-DL-Arg-OH.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1075806

  SDS
alfa aesar™
A1075806
5g Each for $35.55
Description & Specifications

Specifications

Formula Weight 228.69 (210.67 Anhydrous)
Synonym DL-2-Amino-5-guanidinopentanoic acid monohydrochloride monohydrate; H-DL-Arg-OH.HCl
Quantity 5g
Chemical Name or Material DL-Arginine monohydrochloride monohydrate
MDL Number MFCD00151031
TSCA TSCA
Molecular Formula C6H14N4O2·HCl·H2O
CAS 332360-01-7
Assay ≥98%
Beilstein 4158960