missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Arginine hydrochloride, 98%, anhydrous, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446020050
Specifications
DL-Arginine hydrochloride | |
3% max. | |
200.0°C to 203.0°C | |
32042-43-6 | |
C6H14N4O2·ClH | |
1-hydrochloride arginine, 2-amino-5-guanidinopentanoic acid hydrochloride, arginine monochloride, argininhydrochlorid, dl-+/--arginine hydrochloride, dl-arginine hcl, dl-arginine hydrochloride, h-dl-arg-oh.hcl, h-dl-arg-oh.xhcl, wln: muyzm3yzvq &gh | |
KWTQSFXGGICVPE-UHFFFAOYSA-N | |
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride | |
85880 | |
Powder |
Authentic | |
White | |
5g | |
97.5 to 102.5% | |
H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl | |
Solubility in water: soluble. | |
C(CC(C(=O)O)N)CN=C(N)N.Cl | |
210.67 | |
210.67 | |
98% |
Chemical Identifiers
32042-43-6 | |
210.67 | |
1-hydrochloride arginine, 2-amino-5-guanidinopentanoic acid hydrochloride, arginine monochloride, argininhydrochlorid, dl-+/--arginine hydrochloride, dl-arginine hcl, dl-arginine hydrochloride, h-dl-arg-oh.hcl, h-dl-arg-oh.xhcl, wln: muyzm3yzvq &gh | |
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride |
C6H14N4O2·ClH | |
KWTQSFXGGICVPE-UHFFFAOYSA-N | |
85880 | |
C(CC(C(=O)O)N)CN=C(N)N.Cl |
Safety and Handling
EINECSNumber : 250-903-