missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Arginine hydrochloride, 98%, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446020010

 View more versions of this product

Catalog No. AC446020010



Specifications

Specifications

DL-Arginine hydrochloride
3% max.
White
1g
97.5 to 102.5%
H2NC(=NH)NH(CH2)3CH(NH2)CO2H·HCl
Solubility in water: soluble.
C(CC(C(=O)O)N)CN=C(N)N.Cl
210.67
210.67
98%
Authentic
Glass Bottle
200.0°C to 203.0°C
32042-43-6
C6H14N4O2·ClH
1-hydrochloride arginine, 2-amino-5-guanidinopentanoic acid hydrochloride, arginine monochloride, argininhydrochlorid, dl-+/--arginine hydrochloride, dl-arginine hcl, dl-arginine hydrochloride, h-dl-arg-oh.hcl, h-dl-arg-oh.xhcl, wln: muyzm3yzvq &gh
KWTQSFXGGICVPE-UHFFFAOYSA-N
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
85880
Powder

Chemical Identifiers

32042-43-6
210.67
1-hydrochloride arginine, 2-amino-5-guanidinopentanoic acid hydrochloride, arginine monochloride, argininhydrochlorid, dl-+/--arginine hydrochloride, dl-arginine hcl, dl-arginine hydrochloride, h-dl-arg-oh.hcl, h-dl-arg-oh.xhcl, wln: muyzm3yzvq &gh
2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
C6H14N4O2·ClH
KWTQSFXGGICVPE-UHFFFAOYSA-N
85880
C(CC(C(=O)O)N)CN=C(N)N.Cl
Safety and Handling

Safety and Handling

EINECSNumber : 250-903-

SDS Documents