Please login to your online account to display your discounted pricing

DL-AP7, Tocris

Specific NMDA antagonist

$146.74 - $574.46

Chemical Identifiers

Chemical Name or Material DL-AP7
Assay Percent Range >98%
Molecular Formula C7H16NO5P
Formula Weight 225.18
Synonym DL-2-Amino-7-phosphonoheptanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

010450

 
r & d systems
RDS010450
50mg Each for $574.46

010410

 
r & d systems
RDS010410
10mg Each for $146.74
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 225.18
CAS 78966-69-5
Molecular Formula C7H16NO5P
Chemical Name or Material DL-AP7
Assay Percent Range >98%
Synonym DL-2-Amino-7-phosphonoheptanoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature