Please login to your online account to display your discounted pricing

DL-AP7, Tocris


Specific NMDA antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS010450

 View more versions of this product

Catalog No. 010450

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 225.18
CAS 78966-69-5
Chemical Name or Material DL-AP7
Molecular Formula C7H16NO5P
Assay Percent Range >98%
Synonym DL-2-Amino-7-phosphonoheptanoic acid
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material DL-AP7
Assay Percent Range >98%
Molecular Formula C7H16NO5P
Formula Weight 225.18
Synonym DL-2-Amino-7-phosphonoheptanoic acid
Accessories

Store at Room Temperature