Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-alpha-(Methylaminomethyl)benzyl alcohol, 99%

$42.01 - $145.77

Chemical Identifiers

Chemical Name or Material DL-alpha-(Methylaminomethyl)benzyl alcohol
CAS 6589-55-5
Molecular Formula C9H13NO
Formula Weight 151.21
MDL Number MFCD00004506
Beilstein 1072841
Merck Index 14,4601
Synonym 2-Methylamino-1-phenylethanol; Halostachine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0811104

 
alfa aesar™
L0811104
2g Each for $42.01

AAL0811109

 
alfa aesar™
L0811109
10g Each for $145.77
Description & Specifications

Specifications

Formula Weight 151.21
Chemical Name or Material DL-alpha-(Methylaminomethyl)benzyl alcohol
Assay 99%
Molecular Formula C9H13NO
MDL Number MFCD00004506
Beilstein 1072841
Merck Index 14,4601
Synonym 2-Methylamino-1-phenylethanol; Halostachine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 73° to 76°C
Quantity 2g
CAS 6589-55-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DO9625000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.