missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Allothreonine (contains Threonine) 98.0+%, TCI America™

$99.31 - $435.42

Chemical Identifiers

CAS 144-98-9
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00067248
InChI Key AYFVYJQAPQTCCC-HRFVKAFMSA-N
Synonym l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
PubChem CID 99289
ChEBI CHEBI:28718
IUPAC Name (2S,3S)-2-amino-3-hydroxybutanoic acid
SMILES CC(C(C(=O)O)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A0214100MG View Documents TCI America
A0214100MG
100mg Each for $99.31
 
A02141G View Documents TCI America
A02141G
1g Each for $435.42
 
Specifications

Chemical Identifiers

144-98-9
119.12
AYFVYJQAPQTCCC-HRFVKAFMSA-N
99289
(2S,3S)-2-amino-3-hydroxybutanoic acid
C4H9NO3
MFCD00067248
l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
CHEBI:28718
CC(C(C(=O)O)N)O

Specifications

White
144-98-9
MFCD00067248
AYFVYJQAPQTCCC-HRFVKAFMSA-N
(2S,3S)-2-amino-3-hydroxybutanoic acid
99289
119.12
≥98.0% (T)
100mg
C4H9NO3
l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
CC(C(C(=O)O)N)O
119.12
CHEBI:28718
Crystal-Powder at 20°C
DL-Allothreonine (contains Threonine)
Safety and Handling

Safety and Handling

TSCA : No