Please login to your online account to display your discounted pricing

DL-Alanine methyl ester hydrochloride, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393310250

 View more versions of this product

Catalog No. AC393310250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Powder
Formula Weight 139.58
Quantity 25g
Color White to off-white
Packaging Glass bottle
Molecular Formula C4H9NO2·HCl
CAS 13515-97-4
Chemical Name or Material DL-Alanine methyl ester hydrochloride
Assay Percent Range 98+%
Health Hazard 1 Warning
MDL Number MFCD00035523
Linear Formula CH3CH(NH2)COOCH3·HCl
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material DL-Alanine methyl ester hydrochloride
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H9NO2·HCl
Linear Formula CH3CH(NH2)COOCH3·HCl
Formula Weight 139.58
MDL Number MFCD00035523
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents