missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Alanine Ethyl Ester Hydrochloride 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC400430050

 View more versions of this product

Catalog No. AC400430050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

DL-Alanine ethyl ester hydrochloride
617-27-6
Glass bottle
≥98.5%
C5H11NO2·HCl
153.61
Ethyl-2-aminopropanoate hydrochloride
5g
99%
Authentic
Granular Powder and Granules
99%
CH3CH(NH)2COOC2H5·HCl
MFCD00013018
White

Chemical Identifiers

617-27-6
C5H11NO2·HCl
153.61
Ethyl-2-aminopropanoate hydrochloride
≥98.5%
99%
CH3CH(NH)2COOC2H5·HCl
MFCD00013018
DL-Alanine ethyl ester hydrochloride, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 210-507-0

TSCA : TSCA