Please login to your online account to display your discounted pricing

DL-Alanine Ethyl Ester Hydrochloride 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material DL-Alanine ethyl ester hydrochloride
Name Note 99%
CAS 617-27-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H11NO2·HCl
Linear Formula CH3CH(NH)2COOC2H5·HCl
Formula Weight 153.61
MDL Number MFCD00013018
Synonym Ethyl-2-aminopropanoate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC400430050

  SDS
thermo fisher scientific
AC400430050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC400430250

  SDS
fisher scientific acros
400430250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 153.61
Physical Form Granular Powder and Granules
Quantity 5g
Color White
EINECS Number 210-507-0
TSCA TSCA
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 617-27-6
Chemical Name or Material DL-Alanine ethyl ester hydrochloride
Assay Percent Range 99%
Linear Formula CH3CH(NH)2COOC2H5·HCl
MDL Number MFCD00013018
Name Note 99%
Molecular Formula C5H11NO2·HCl
Synonym Ethyl-2-aminopropanoate hydrochloride
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 HYGROSCOPIC
Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.