Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Alanine ethyl ester hydrochloride, 99%

$51.38 - $198.89

Chemical Identifiers

Chemical Name or Material DL-Alanine ethyl ester hydrochloride
CAS 617-27-6
Molecular Formula C5H11NO2·HCl
Formula Weight 153.61
MDL Number MFCD00013018
Beilstein 3654425
Synonym H-DL-Ala-OEt·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1102006

  SDS
alfa aesar™
L1102006
5g Each for $51.38

AAL1102014

  SDS
alfa aesar™
L1102014
25g Each for $198.89
Description & Specifications

Specifications

Formula Weight 153.61
Chemical Name or Material DL-Alanine ethyl ester hydrochloride
Assay 99%
Synonym H-DL-Ala-OEt·HCl
MDL Number MFCD00013018
Molecular Formula C5H11NO2·HCl
TSCA TSCA
Beilstein 3654425
Quantity 5g
Melting Point 84° to 86°C
CAS 617-27-6