missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Alanine ethyl ester hydrochloride, 99%

Manufacturer:  Alfa Aesar™ L1102014

 View more versions of this product

Catalog No. AAL1102014


 

Specifications Description & Specifications

Specifications

153.61
3654425
25g
H-DL-Ala-OEt·HCl
DL-Alanine ethyl ester hydrochloride
99%
84° to 86°C
617-27-6
MFCD00013018
C5H11NO2·HCl

Chemical Identifiers

617-27-6
153.61
3654425
DL-Alanine ethyl ester hydrochloride
C5H11NO2·HCl
MFCD00013018
H-DL-Ala-OEt·HCl

Safety and Handling

SDS & Safety Documents

TSCA : TSCA