missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Alanine ethyl ester hydrochloride, 99%

Manufacturer:  Alfa Aesar™ L1102006

 View more versions of this product

Catalog No. AAL1102006



Specifications

Specifications

DL-Alanine ethyl ester hydrochloride
C5H12ClNO2
3654425
JCXLZWMDXJFOOI-UHFFFAOYSA-N
ethyl 2-aminopropanoate;hydrochloride
102560
99%
5g
617-27-6
MFCD00013018
alanine, ethyl ester, hydrochloride
CCOC(=O)C(C)N.Cl
153.606
153.61
84°C to 86°C

Chemical Identifiers

617-27-6
153.606
JCXLZWMDXJFOOI-UHFFFAOYSA-N
102560
CCOC(=O)C(C)N.Cl
C5H12ClNO2
MFCD00013018
alanine, ethyl ester, hydrochloride
ethyl 2-aminopropanoate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents