missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Alanine 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 159095000

 View more versions of this product

Catalog No. AC159095000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

DL-Alanine
200ppm max.
1ppm max.
99%
C3H7NO2
Authentic
10ppm max.
0.2% max. (105°C, 3 hrs)
0.5% max. (TLC)
Plastic bottle
CC(C(=O)O)N
89.094
CHEBI:16449
Crystals or Crystalline Powder
300ppm max.
95% min. (c=10, H2O, 430nm) 1cm cell
>190 °C
272.0°C to 275.0°C
500g
99%
302-72-7
200ppm max.
10ppm max.
CH3CH(NH2)CO2H
MFCD00064408
04, 387
15, 197
+--alanine
QNAYBMKLOCPYGJ-UHFFFAOYSA-N
2-aminopropanoic acid
602
89.09
Solubility in water: 156g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents, soluble in mineral acids, soluble in ethanol 0.0087g/100g (25°C)
99%
0.1% max.
White
5.5 to 7 (2.5% soln. at 25°C)

Chemical Identifiers

302-72-7
89.094
QNAYBMKLOCPYGJ-UHFFFAOYSA-N
602
2-aminopropanoic acid
C3H7NO2
MFCD00064408
+--alanine
CHEBI:16449
CC(C(=O)O)N
Safety and Handling

Safety and Handling

EINECSNumber : 206-126-4

RTECSNumber : AY2980000

TSCA : TSCA

SDS Documents