missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Adrenaline

A hormone and neurotransmitter

Manufacturer:  Alfa Aesar™ J6300514

 View more versions of this product

Catalog No. AAJ6300514



Specifications

Specifications

Light sensitive
197°C (decomposition)
329-65-7
(HO)2C6H3CH(CH2NHCH3)OH
UN2811
+--adrenaline, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol, dl-adrenaline, dl-epinephrine, epinephrine racemic, racepinefrina, racepinefrine, racepinefrinum, racepinephrine
CNCC(C1=CC(=C(C=C1)O)O)O
183.207
CHEBI:33568
Powder
White
25g
C9H13NO3
MFCD00063027
14,3619
UCTWMZQNUQWSLP-UHFFFAOYSA-N
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
838
183.2

Chemical Identifiers

329-65-7
183.207
UCTWMZQNUQWSLP-UHFFFAOYSA-N
838
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
C9H13NO3
MFCD00063027
+--adrenaline, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol, dl-adrenaline, dl-epinephrine, epinephrine racemic, racepinefrina, racepinefrine, racepinefrinum, racepinephrine
CHEBI:33568
CNCC(C1=CC(=C(C=C1)O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H311-H315-H319-H335
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P501c

H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 206-347-6

RTECSNumber : DO2975000

TSCA : No

Recommended Storage : Keep cold

Documents