missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-Adrenaline

A hormone and neurotransmitter

Manufacturer:  Alfa Aesar™ J6300506

 View more versions of this product

Catalog No. AAJ6300506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

DL-Adrenaline
White
5g
C9H13NO3
MFCD00063027
14,3619
UCTWMZQNUQWSLP-UHFFFAOYSA-N
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
838
183.2
Light Sensitive
197°C
329-65-7
(HO)2C6H3CH(CH2NHCH3)OH
UN2811
dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
CNCC(C1=CC(=C(C=C1)O)O)O
183.207
CHEBI:33568
Powder

Chemical Identifiers

329-65-7
183.207
UCTWMZQNUQWSLP-UHFFFAOYSA-N
838
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
C9H13NO3
MFCD00063027
dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol
CHEBI:33568
CNCC(C1=CC(=C(C=C1)O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H311-H315-H319-H335
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DO2975000

Recommended Storage : 2° to 8°C

Documents