Learn More
DL-Adrenaline, Thermo Scientific Chemicals
A hormone and neurotransmitter
Supplier: Thermo Scientific Chemicals J6300503
Description
A hormone and neurotransmitter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-Adrenaline | |
197°C (decomposition) | |
C9H13NO3 | |
MFCD00063027 | |
UN2811 | |
14,3619 | |
UCTWMZQNUQWSLP-UHFFFAOYSA-N | |
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol | |
838 | |
183.2 |
329-65-7 | |
White | |
(HO)2C6H3CH(CH2NHCH3)OH | |
1 g | |
Light sensitive | |
dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol | |
CNCC(C1=CC(=C(C=C1)O)O)O | |
183.207 | |
CHEBI:33568 | |
Powder |
Chemical Identifiers
329-65-7 | |
183.207 | |
UCTWMZQNUQWSLP-UHFFFAOYSA-N | |
838 | |
4-[1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
C9H13NO3 | |
MFCD00063027 | |
dl-adrenaline, racepinephrine, racepinefrine, dl-epinephrine, +--adrenaline, epinephrine racemic, racepinefrina, racepinefrinum, +--epinephrine, 4-1-hydroxy-2-methylamino ethyl benzene-1,2-diol | |
CHEBI:33568 | |
CNCC(C1=CC(=C(C=C1)O)O)O |
Safety and Handling
GHS H Statement
H311-H315-H319-H335
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P501c
H311-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 206-347-6
RTECSNumber : DO2975000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only