Please login to your online account to display your discounted pricing

DL-6-Methyl-5-hepten-2-ol, 99%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC187691000

 View more versions of this product

Catalog No. AC187691000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 128.21
Physical Form Liquid
Quantity 100mL
Color Clear colorless to slightly yellow
Density 0.84g/mL
Beilstein 01, IV, 2167
CAS 997266
Packaging Glass bottle
Chemical Name or Material DL-6-Methyl-5-hepten-2-ol
Molecular Formula C8H16O
Assay Percent Range 99%
MDL Number MFCD00004561
Infrared Spectrum Authentic
Linear Formula (CH3)2C=CHCH2CH2CH(OH)CH3
Refractive Index 1.4475 to 1.4495 (20°C, 589nm)

Specifications

Chemical Name or Material DL-6-Methyl-5-hepten-2-ol
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H16O
Linear Formula (CH3)2C=CHCH2CH2CH(OH)CH3
Formula Weight 128.21
MDL Number MFCD00004561