missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-6-Methoxy-α-methyl-2-naphthalenemethanol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 331500250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-6-Methoxy-α-methyl-2-naphthalenemethanol | |
77301-42-9 | |
Beige-Yellow to White | |
97.5% min. (HPLC) | |
C13H14O2 | |
25 g | |
Solubility in water: insoluble. Other solubilities: soluble in acetone,dichloromethane,ethylacetate,slightly soluble in benzene | |
COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O | |
202.25 | |
202.25 | |
Crystalline Powder |
98% | |
111.0°C to 114.0°C | |
Authentic | |
Glass bottle | |
MFCD01632581 | |
1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k | |
OUVJWFRUESFCCY-SECBINFHSA-N | |
1-(6-methoxynaphthalen-2-yl)ethanol | |
575523 | |
98% |
Chemical Identifiers
77301-42-9 | |
202.25 | |
OUVJWFRUESFCCY-SECBINFHSA-N | |
575523 | |
COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O |
C13H14O2 | |
MFCD01632581 | |
1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k | |
1-(6-methoxynaphthalen-2-yl)ethanol |
RUO – Research Use Only