Please login to your online account to display your discounted pricing

DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 97%, ACROS Organics™


Quantity 250mg
Packaging Glass Bottle in Plastic Container

Manufacturer: thermo fisher scientific  326082500

 View more versions of this product

Catalog No. AC326082500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 191
Quantity 250mg
Color Light beige
Packaging Glass Bottle in Plastic Container
Infrared Spectrum Authentic
Chemical Name or Material DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Name Note 97%
Assay Percent Range 97%
Molecular Formula C11 H13 N O2
MDL Number MFCD01320673
CAS 151637-59-1

Specifications

Chemical Name or Material DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Name Note 97%
CAS 151637-59-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11 H13 N O2
Formula Weight 191
MDL Number MFCD01320673