Please login to your online account to display your discounted pricing

DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  326080010

 View more versions of this product

Catalog No. AC326080010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 191
Physical Form Powder
Quantity 1g
Color Light beige
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
MDL Number MFCD01320673
CAS 151637-59-1
Name Note 97%
Molecular Formula C11 H13 N O2
Assay Percent Range 97%

Specifications

Chemical Name or Material DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Name Note 97%
CAS 151637-59-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11 H13 N O2
Formula Weight 191
MDL Number MFCD01320673