Please login to your online account to display your discounted pricing

DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 97%, ACROS Organics™

$171.46 - $171.46

Chemical Identifiers

Chemical Name or Material DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Name Note 97%
CAS 151637-59-1
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11 H13 N O2
Formula Weight 191
MDL Number MFCD01320673
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC326080010

  SDS
thermo fisher scientific
326080010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC326082500

  SDS
thermo fisher scientific
326082500
Glass Bottle in Plastic Container 250mg Each for $171.46
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 191
Physical Form Powder
Quantity 1g
Color Light beige
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material DL-5-Methyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
MDL Number MFCD01320673
CAS 151637-59-1
Name Note 97%
Molecular Formula C11 H13 N O2
Assay Percent Range 97%