Please login to your online account to display your discounted pricing

DL-5-Methoxytryptophan, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  151760050

 View more versions of this product

Catalog No. AC151760050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Appearance of Solution (1% soln. in warm water) clear, colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Water 3% max. (K.F.)
Color Off-white
Quantity 5g
Chemical Name or Material DL-5-Methoxytryptophan
CAS 28052-84-8
Physical Form Off-white powder
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C12H14N2O3
Formula Weight 234.25g/mol
MDL Number MFCD00005650

Specifications

Chemical Name or Material DL-5-Methoxytryptophan
CAS 28052-84-8
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C12H14N2O3
Formula Weight 234.25g/mol
MDL Number MFCD00005650