Please login to your online account to display your discounted pricing

DL-5-Methoxytryptophan, 95%, ACROS Organics™

$71.64 - $385.72

Chemical Identifiers

Chemical Name or Material DL-5-Methoxytryptophan
CAS 28052-84-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H14N2O3
Formula Weight 234.25
MDL Number MFCD00005650
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC151760050

  SDS
thermo fisher scientific
AC151760050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC151760010

  SDS
thermo fisher scientific
151760010
Glass bottle 1g Each for $385.72

AC151761000

  SDS
thermo fisher scientific
151761000
Glass bottle 100mg Each for $71.64
Description & Specifications

Specifications

Physical Form Powder
Assay ≥94%
Formula Weight 234.25
Quantity 5g
Color Off-white
Chemical Name or Material DL-5-Methoxytryptophan
Infrared Spectrum Authentic
Molecular Formula C12H14N2O3
Water 3% max. (K.F.)
Appearance of Solution (1% soln. in warm water) clear, colorless
MDL Number MFCD00005650
CAS 28052-84-8
Packaging Glass bottle
Assay Percent Range 95%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.