missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-5-Methoxytryptophan, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 151761000

 View more versions of this product

Catalog No. AC151761000


 

Specifications Description & Specifications

Specifications

DL-5-Methoxytryptophan
(1% soln. in warm water) clear, colorless
≥94%
Authentic
234.25
Glass bottle
Off-white
28052-84-8
Powder
95%
C12H14N2O3
MFCD00005650
3% max. (K.F.)
100mg

Chemical Identifiers

28052-84-8
C12H14N2O3
MFCD00005650
≥94%
95%
234.25
DL-5-Methoxytryptophan