Please login to your online account to display your discounted pricing

DL-5-Methoxytryptophan, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  151760010

 View more versions of this product

Catalog No. AC151760010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 234.25
Assay ≥94%
Quantity 1g
Color Off-white
Appearance of Solution (1% soln. in warm water) clear, colorless
CAS 28052-84-8
Chemical Name or Material DL-5-Methoxytryptophan
Molecular Formula C12H14N2O3
MDL Number MFCD00005650
Packaging Glass bottle
Infrared Spectrum Authentic
Water 3% max. (K.F.)
Assay Percent Range 95%

Specifications

Chemical Name or Material DL-5-Methoxytryptophan
CAS 28052-84-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H14N2O3
Formula Weight 234.25
MDL Number MFCD00005650