Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-5-Hydroxytryptophan, 99%

$81.12 - $81.12

Chemical Identifiers

Chemical Name or Material DL-5-Hydroxytryptophan
CAS 56-69-9
Molecular Formula C11H12N2O3
Formula Weight 220.23
MDL Number MFCD00005651
Beilstein 88199
Merck Index 14,4847
Synonym DL-2-Amino-3-[3-(5-hydroxyindole)]propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1223703

 
alfa aesar™
A1223703
1g Each for $81.12
Description & Specifications

Specifications

Formula Weight 220.23
Chemical Name or Material DL-5-Hydroxytryptophan
TSCA TSCA
Assay 99%
Molecular Formula C11H12N2O3
MDL Number MFCD00005651
CAS 56-69-9
Beilstein 88199
Merck Index 14,4847
Synonym DL-2-Amino-3-[3-(5-hydroxyindole)]propionic acid
Melting Point ≈298°C (dec.)
Quantity 1g
RTECS Number YN7100000

Applications
It is an immediate precursor that is used to assess central serotonergic function. It was used for the treatment of insomnia.

Solubility
Soluble in water (4 mg/ml at 25°C), methanol.

Notes
Store at 4°C. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.