Please login to your online account to display your discounted pricing

DL-5-Benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  339850010

 View more versions of this product

Catalog No. AC339850010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 277.32
Physical Form Fine Crystalline Powder and Chunks
Quantity 1g
Color White
Chemical Name or Material DL-5-Benzoylamino-5-isobutyl-4-oxo-1, 3-dioxane
Assay Percent Range 95%
CAS 51127-26-5
Molecular Formula C15 H19 N O4
MDL Number MFCD01863312
Name Note 95%
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material DL-5-Benzoylamino-5-isobutyl-4-oxo-1, 3-dioxane
Name Note 95%
CAS 51127-26-5
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C15 H19 N O4
Formula Weight 277.32
MDL Number MFCD01863312