missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
DL-5-Benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, 95%, ACROS Organics™
Chemical Identifiers
CAS |
51127-26-5 |
Molecular Formula |
C15H19NO4 |
Molecular Weight (g/mol) |
277.32 |
MDL Number |
MFCD01863312 |
InChI Key |
ZJPYVGCXULFGJA-UHFFFAOYSA-N |
Synonym |
5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,95, dl-5-benzoylamino-5-isobutyl-4-oxo-1?3-dioxane, n-5-2-methyl-propyl-4-oxo-1,3 dioxan-5-yl-benzamide, n-5-2-methylpropyl-4-oxo 1,3-dioxan-5-yl benzamide, n-5-2-methylpropyl-4-oxo-1,3-dioxan-5-yl benzamide |
PubChem CID |
2733906 |
IUPAC Name |
N-[5-(2-methylpropyl)-4-oxo-1,3-dioxan-5-yl]benzamide |
SMILES |
CC(C)CC1(COCOC1=O)NC(=O)C2=CC=CC=C2 |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
51127-26-5 |
|
277.32 |
|
ZJPYVGCXULFGJA-UHFFFAOYSA-N |
|
2733906 |
|
CC(C)CC1(COCOC1=O)NC(=O)C2=CC=CC=C2 |
|
C15H19NO4 |
|
MFCD01863312 |
|
5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,95, dl-5-benzoylamino-5-isobutyl-4-oxo-1?3-dioxane, n-5-2-methyl-propyl-4-oxo-1,3 dioxan-5-yl-benzamide, n-5-2-methylpropyl-4-oxo 1,3-dioxan-5-yl benzamide, n-5-2-methylpropyl-4-oxo-1,3-dioxan-5-yl benzamide |
|
N-[5-(2-methylpropyl)-4-oxo-1,3-dioxan-5-yl]benzamide |
Specifications
|
Authentic |
|
White |
|
51127-26-5 |
|
C15H19NO4 |
|
5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane, dl-5-benzoylamino-5-isobutyl-4-oxo-1,3-dioxane,95, dl-5-benzoylamino-5-isobutyl-4-oxo-1?3-dioxane, n-5-2-methyl-propyl-4-oxo-1,3 dioxan-5-yl-benzamide, n-5-2-methylpropyl-4-oxo 1,3-dioxan-5-yl benzamide, n-5-2-methylpropyl-4-oxo-1,3-dioxan-5-yl benzamide |
|
CC(C)CC1(COCOC1=O)NC(=O)C2=CC=CC=C2 |
|
277.32 |
|
277.32 |
|
≥94% |
|
Glass Bottle |
|
1g |
|
95% |
|
MFCD01863312 |
|
ZJPYVGCXULFGJA-UHFFFAOYSA-N |
|
N-[5-(2-methylpropyl)-4-oxo-1,3-dioxan-5-yl]benzamide |
|
2733906 |
|
Fine Crystalline Powder and Chunks |
|
DL-5-Benzoylamino-5-isobutyl-4-oxo-1, 3-dioxane, 95% |
SDS