Please login to your online account to display your discounted pricing

Alfa Aesar™ DL-4-Hydroxymandelic acid monohydrate, 97%

$36.01 - $36.01

Chemical Identifiers

Chemical Name or Material DL-4-Hydroxymandelic acid monohydrate
CAS 184901-84-6
Molecular Formula C8H8O4·H2O
Formula Weight 186.17 (168.15 Anhydrous)
MDL Number MFCD00149283
Beilstein 2365374
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0591606

  SDS
alfa aesar™
L0591606
5g Each for $36.01
Description & Specifications

Specifications

Formula Weight 186.17 (168.15 Anhydrous)
Chemical Name or Material DL-4-Hydroxymandelic acid monohydrate
Assay 97%
MDL Number MFCD00149283
CAS 184901-84-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Molecular Formula C8H8O4·H2O
Beilstein 2365374
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Recommended Storage 2° to 8°C
Melting Point 88° to 90°C
Quantity 5g

Applications
DL-4-Hydroxymandelic acid monohydrate is used in the synthesis of an enzyme activity-based labeling molule.

Solubility
Soluble in water.

Notes
Incompatible with air, heat and oxidizing agents. Store away from air. Protect from heat.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.

2° to 8°C